Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000268.3 | NP_000259.1 | 2 | 578 | ||
NM_016418.5 | NP_057502.2 | 2 | 578 | |||
NM_181825.2 | NP_861546.1 | 2 | 578 | |||
NM_181829.2 | NP_861967.1 | 2 | 578 | |||
NM_181832.2 | NP_861970.1 | 2 | 578 | |||
NM_181833.2 | NP_861971.1 | 2 | 578 | |||
XM_017028809.1 | XP_016884298.1 | 2 | 141 | |||
XM_017028810.1 | XP_016884299.1 | 2 | 137 | |||
GenBank mRNA | AF122827.1 | - | 2 | 185 | ||
AF123570.1 | - | 2 | 185 | |||
AF369657.1 | - | 2 | 560 | |||
AF369658.1 | - | 2 | 560 | |||
AF369661.1 | - | 2 | 135 | |||
AF369662.1 | - | 2 | 135 | |||
AF369664.1 | - | 2 | 135 | |||
AF369666.1 | - | 3 | 251 | |||
AF369667.1 | - | 2 | 135 | |||
AF369668.1 | - | 2 | 135 | |||
AF369701.1 | - | 2 | 501 | |||
AK297116.1 | - | 2 | 517 | |||
BC020257.1 | - | 2 | 473 | 4871980 | ||
CF138178.1 | - | 2 | 359 | |||
CR456530.1 | - | 2 | 197 | |||
L11353.1 | - | 2 | 354 | |||
Z22664.1 | - | 2 | 309 |