Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026535.2 | NP_080811.1 | 2 | 731 | ||
GenBank mRNA | AK014346.1 | - | 2 | 673 | ||
AK014589.1 | - | 2 | 731 | |||
AY326419.1 | - | 1 | 564 | |||
BC062143.1 | - | 2 | 685 | 30287005 |