Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnaseh2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026616.2 | NP_080892.1 | 4 | 539 | ||
GenBank mRNA | AF346604.1 | - | 4 | 537 | ||
AK011556.1 | - | 4 | 517 | |||
AK018776.1 | - | 4 | 528 | |||
BC024333.1 | - | 4 | 511 | 5040687 | ||
CD553809.1 | - | 1 | 30 |