Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026752.4 | NP_081028.3 | 1, 2 | 213 | ||
GenBank mRNA | AK003661.1 | - | 1, 2 | 110 | ||
AK139356.1 | - | 148 | ||||
AK155300.1 | - | 1, 2 | 214 | |||
AK171099.1 | - | 1, 2 | 154 | |||
BC019521.1 | - | 1, 2 | 143 | 4220205 | ||
BC132248.1 | - | 1, 2 | 113 | 40130525 | ||
BC132250.1 | - | 1, 2 | 113 | 40130527 |