Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Urm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026615.4 | NP_080891.1 | 5 | 474 | ||
GenBank mRNA | AK012124.1 | - | 5 | 480 | ||
AK019352.1 | - | 5 | 480 | |||
AK049227.1 | - | 5 | 475 | |||
AK049849.1 | - | 5 | 475 | |||
AK165008.1 | - | 2 | 1474 | |||
BC026994.1 | - | 5 | 466 | 5368627 | ||
BC060255.1 | - | 2 | 1347 |