Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026853.1 | NP_081129.1 | 3 | 291 | ||
GenBank mRNA | AF398972.1 | - | 3 | 405 | ||
AK002412.1 | - | 3 | 378 | |||
AK004396.1 | - | 3 | 291 | |||
AK005404.1 | - | 3 | 405 | |||
AK146664.1 | - | 3 | 405 | |||
BC064047.1 | - | 3 | 287 | 864428 |