Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039483.1 | NP_001034572.1 | 2 | 297 | ||
GenBank mRNA | AK004504.1 | - | 2 | 139 | ||
AK015068.1 | - | 2 | 588 | |||
AK037709.1 | - | 2 | 232 | |||
AK162344.1 | - | 2 | 236 | |||
AK183030.1 | - | 2 | 50 | |||
AK218449.1 | - | 2 | 50 | |||
BC011457.1 | - | 2 | 171 | 3585288 | ||
BC019761.1 | - | 2 | 215 | 5009960 | ||
BC020098.1 | - | 2 | 227 | 4019484 |