Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebna1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026932.4 | NP_081208.1 | 2 | 244 | ||
GenBank mRNA | AK007491.1 | - | 2 | 214 | ||
AK134732.1 | - | 2 | 199 | |||
AK167070.1 | - | 2 | 199 | |||
AK168269.1 | - | 2 | 171 | |||
BC054723.1 | - | 2 | 227 | 5695176 | ||
BC062876.1 | - | 2 | 176 | 5686021 | ||
BY324791.1 | - | 2 | 244 |