Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a6d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026835.2 | NP_081111.1 | 7 | 824 | ||
GenBank mRNA | AF237911.1 | - | 7 | 786 | ||
AK004295.1 | - | 7 | 807 | |||
AK152859.1 | - | 7 | 805 | |||
AK156878.1 | - | 7 | 807 | |||
AY039037.1 | - | 7 | 786 | |||
BC018331.1 | - | 7 | 761 | 3964037 | ||
BC029738.1 | - | 7 | 768 | 4016611 |