Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000197.1 | NP_000188.1 | 4 | 345 | ||
XM_011518618.2 | XP_011516920.1 | 7 | 817 | |||
XM_011518619.2 | XP_011516921.1 | 6 | 655 | |||
XM_017014671.1 | XP_016870160.1 | 6 | 2729 | |||
XM_017014672.1 | XP_016870161.1 | 6 | 2653 | |||
XM_017014673.1 | XP_016870162.1 | 6 | 2730 | |||
XM_017014675.1 | XP_016870164.1 | 5 | 2410 | |||
XM_017014676.1 | XP_016870165.1 | 6 | 2731 | |||
XM_017014677.1 | XP_016870166.1 | 6 | 2734 | |||
GenBank mRNA | BC034281.1 | - | 4 | 329 | 5168012 | |
BT019371.1 | - | 4 | 297 | |||
CR541894.1 | - | 4 | 297 | |||
KU178130.1 | - | 4 | 297 | |||
KU178131.1 | - | 4 | 297 | |||
KU178132.1 | - | 4 | 297 | |||
KU178133.1 | - | 4 | 297 | |||
U05659.1 | - | 4 | 345 |