Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000212.2 | NP_000203.2 | 4 | 430 | ||
GenBank mRNA | AY826979.1 | - | 4 | 419 | ||
BC127666.1 | - | 4 | 430 | 40128462 | ||
BC127667.1 | - | 4 | 434 | 40128464 | ||
J02703.1 | - | 4 | 430 | |||
KC120777.1 | - | 4 | 430 | |||
KC120778.1 | - | 4 | 430 | |||
M20311.1 | - | 4 | 426 | |||
M35999.1 | - | 4 | 428 | |||
U95204.1 | - | 4 | 410 |