Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027052.3 | NP_081328.2 | 5, 6 | 522 | ||
GenBank mRNA | AB055004.1 | - | 5, 6 | 498 | ||
AK051023.1 | - | 5, 6 | 523 | |||
AK164292.1 | - | 5, 6 | 465 | |||
AK167539.1 | - | 5, 6 | 504 | |||
AK169349.1 | - | 4, 5 | 511 | |||
AY027919.1 | - | 5, 6 | 561 | |||
BC024072.1 | - | 5, 6 | 447 | 5097293 | ||
BC024123.1 | - | 5, 6 | 479 | 5098067 | ||
BC031717.1 | - | 5, 6 | 440 | 4195569 |