Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000226.3 | NP_000217.2 | 1 | 360 | ||
GenBank mRNA | S69510.1 | - | 1 | 357 | ||
Z29074.1 | - | 1 | 360 |