Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000244.3 | NP_000235.2 | 4 | 806 | ||
NM_130799.2 | NP_570711.1 | 4 | 791 | |||
NM_130800.2 | NP_570712.1 | 4 | 769 | |||
NM_130801.2 | NP_570713.1 | 4 | 757 | |||
NM_130802.2 | NP_570714.1 | 4 | 1193 | |||
NM_130803.2 | NP_570715.1 | 4 | 1200 | |||
NM_130804.2 | NP_570716.1 | 5 | 1072 | |||
XM_005274001.4 | XP_005274058.1 | 4 | 1173 | |||
XM_011545040.1 | XP_011543342.1 | 4 | 1173 | |||
XM_011545041.2 | XP_011543343.1 | 4 | 872 | |||
XM_011545042.2 | XP_011543344.1 | 4 | 777 | |||
XM_017017765.1 | XP_016873254.1 | 4 | 1188 | |||
XM_017017766.1 | XP_016873255.1 | 4 | 887 | |||
XM_017017767.1 | XP_016873256.1 | 4 | 794 | |||
XM_017017768.1 | XP_016873257.1 | 4 | 819 | |||
XM_017017769.1 | XP_016873258.1 | 4 | 872 | |||
XM_017017770.1 | XP_016873259.1 | 4 | 775 | |||
GenBank mRNA | BC002544.2 | - | 4 | 644 | 3138471 | |
U93236.1 | - | 4 | 791 |