Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027276.2 | NP_081552.2 | 14, 15 | 1439 | ||
GenBank mRNA | AK013213.1 | - | 3, 4 | 373 | ||
AK147938.1 | - | 14, 15 | 1454 | |||
AK167965.1 | - | 14, 15 | 1454 | |||
AK168978.1 | - | 14, 15 | 1426 | |||
AK188605.1 | - | 16 | ||||
AK201828.1 | - | 2, 3 | 158 | |||
BC026606.1 | - | 14, 15 | 1439 | 4976424 |