Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027290.3 | NP_081566.2 | 3 | 421 | ||
GenBank mRNA | AK021211.1 | - | 2 | 345 | ||
AK030780.1 | - | 3 | 366 | |||
AK041406.1 | - | 3 | 356 | |||
BC064314.1 | - | 3 | 784 | |||
BC094576.1 | - | 3 | 421 | 6853824 | ||
BC120687.1 | - | 2 | 330 | 40129610 | ||
BC120689.1 | - | 2 | 330 | 40129612 |