Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastkd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027123.4 | NP_081399.3 | 2, 3 | 1575 | ||
GenBank mRNA | AK031142.1 | - | 2, 3 | 1562 | ||
AK033189.1 | - | 2, 3 | 1576 | |||
AK036778.1 | - | 2, 3 | 1561 | |||
AK148902.1 | - | 2, 3 | 1566 | |||
BC060688.1 | - | 2, 3 | 1795 | 6409274 |