Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Steap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027399.3 | NP_081675.2 | 3 | 305 | ||
GenBank mRNA | AF297098.1 | - | 2 | 235 | ||
AK010437.1 | - | 3 | 300 | |||
AK144312.1 | - | 3 | 260 | |||
AY029584.1 | - | 3 | 264 | |||
BC061023.1 | - | 3 | 290 | 30304473 | ||
DN176485.1 | - | 3 | 305 |