Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gulp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028450.3 | NP_082726.2 | 4 | 377 | ||
GenBank mRNA | AK014093.1 | - | 3 | 126 | ||
AK017798.1 | - | 4 | 355 | |||
AK028490.1 | - | 3 | 293 | |||
AK036670.1 | - | 4 | 378 | |||
BC032154.1 | - | 4 | 548 | 5068339 | ||
BY713087.1 | - | 3 | 124 | |||
DQ786776.1 | - | 3 | 131 |