Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168223.1 | NP_001161695.1 | 4, 5 | 425 | ||
NM_001168224.1 | NP_001161696.1 | 4, 5 | 537 | |||
NM_027562.4 | NP_081838.1 | 5, 6 | 599 | |||
GenBank mRNA | AB091386.1 | - | 2, 3 | 231 | ||
AK014570.1 | - | 5, 6 | 600 | |||
AK137753.1 | - | 4, 5 | 535 | |||
AY137341.1 | - | 4, 5 | 537 | |||
AY137342.1 | - | 4, 5 | 425 | |||
BC117543.1 | - | 5, 6 | 589 | 40105656 | ||
JF431014.1 | - | 3, 4 | 377 | |||
JF792238.1 | - | 4, 5 | 436 | |||
JF792239.1 | - | 4, 5 | 487 |