Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027439.4 | NP_081715.1 | 2, 3 | 313 | ||
GenBank mRNA | AB192471.2 | - | 2, 3 | 189 | ||
AK017482.1 | - | 2, 3 | 245 | |||
AK029405.1 | - | 2, 3 | 216 | |||
AK076495.1 | - | 2, 3 | 258 | |||
AK151735.1 | - | 2, 3 | 244 | |||
AK168139.1 | - | 2, 3 | 272 | |||
BC014706.1 | - | 2, 3 | 184 | 4457079 | ||
BY784908.1 | - | 2, 3 | 314 |