Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Medag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027519.3 | NP_081795.1 | 5 | 2028 | ||
GenBank mRNA | AK036605.1 | - | 5 | 2029 | ||
AK042557.1 | - | 5 | 1741 | |||
AK047657.1 | - | 5 | 2085 | |||
AK086092.1 | - | 5 | 2039 | |||
AK138268.1 | - | 5 | 1727 | |||
AK160904.1 | - | 5 | 2034 | |||
AK161526.1 | - | 5 | 2037 | |||
AK162725.1 | - | 5 | 2029 | |||
AK163617.1 | - | 5 | 1727 | |||
AK165529.1 | - | 5 | 1825 | |||
BC026658.1 | - | 5 | 1791 | |||
BC037052.1 | - | 1 | 970 | |||
BC116246.1 | - | 5 | 1950 | 40093530 | ||
BC116247.1 | - | 5 | 1948 | 40093536 |