Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000302.3 | NP_000293.2 | 3 | 307 | ||
NM_001316320.1 | NP_001303249.1 | 4 | 448 | |||
GenBank mRNA | AK294688.1 | - | 3 | 249 | ||
AK295254.1 | - | 3 | 251 | |||
AK299150.1 | - | 4 | 381 | |||
AK312243.1 | - | 3 | 240 | |||
AK316285.1 | - | 4 | 384 | |||
AU139472.1 | - | 3 | 286 | |||
BC016657.2 | - | 3 | 211 | 3917351 | ||
DC334246.1 | - | 3 | 307 | |||
M98252.1 | - | 3 | 218 |