Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Raver1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027911.3 | NP_082187.1 | 1, 2 | 238 | ||
GenBank mRNA | AK039928.1 | - | 1, 2 | 250 | ||
AK051444.1 | - | 1, 2 | 250 | |||
AK079191.1 | - | 1, 2 | 243 | |||
AK084478.1 | - | 1, 2 | 250 | |||
AK138596.1 | - | 1, 2 | 250 | |||
AK168436.1 | - | 1, 2 | 259 | |||
BC058271.1 | - | 1, 2 | 238 | 6490833 | ||
BC108392.1 | - | 1, 2 | 237 | 6393833 |
Gene Symbol: Gm38431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004919.1 | - | 1, 2 | 251 | ||
AY275472.1 | - | 1, 2 | 237 |