Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027869.1 | NP_082145.1 | 4 | 396 | ||
GenBank mRNA | AF465249.1 | - | 4 | 395 | ||
AJ507387.1 | - | 4 | 396 | |||
AK144151.1 | - | 399 | ||||
AK149419.1 | - | 4 | 397 | |||
AK152445.1 | - | 4 | 373 | |||
AK161833.1 | - | 4 | 397 | |||
AK165566.1 | - | 4 | 399 | |||
BC055826.1 | - | 4 | 429 | 3498934 |