Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027878.2 | NP_082154.2 | 7 | 847 | ||
GenBank mRNA | AK004552.1 | - | 7 | 847 | ||
AK151184.1 | - | 7 | 780 | |||
BC021433.1 | - | 7 | 813 | 5030012 |