Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027669.3 | NP_081945.1 | 8 | 963 | ||
GenBank mRNA | AK016798.1 | - | 8 | 891 | ||
AK077091.1 | - | 8 | 964 | |||
BC115618.1 | - | 8 | 874 | 40102763 | ||
BC115619.1 | - | 7 | 1009 | 40102765 |