Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027760.2 | NP_082036.1 | 4 | 1331 | ||
GenBank mRNA | AF424737.1 | - | 4 | 1331 | ||
AK017214.1 | - | 3 | 738 | |||
AK031980.1 | - | 3 | 637 | |||
BC115779.1 | - | 4 | 1302 | 40105476 | ||
BC117546.1 | - | 4 | 1302 | 40105479 |