Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000199.3 | NP_000190.1 | 8 | 2164 | ||
XM_005257582.3 | XP_005257639.1 | 8 | 2273 | |||
XM_011525127.2 | XP_011523429.1 | 9 | 2152 | |||
XM_017024952.1 | XP_016880441.1 | 8 | 3009 | |||
GenBank mRNA | AB209900.1 | - | 5 | 4959 | ||
AK095969.1 | - | 7 | 2018 | |||
BC047318.1 | - | 8 | 2089 | 5226903 | ||
U30894.1 | - | 8 | 2089 |