Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028031.3 | NP_082307.1 | 2 | 254 | ||
GenBank mRNA | AK010382.1 | - | 2 | 254 | ||
AK145855.1 | - | 2 | 174 | |||
AK146196.1 | - | 2 | 226 | |||
BC019493.1 | - | 2 | 173 | |||
BC046599.1 | - | 2 | 173 | 6311721 |