Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000211.4 | NP_000202.3 | 12 | 1738 | ||
NM_001127491.2 | NP_001120963.2 | 12 | 1728 | |||
NM_001303238.1 | NP_001290167.1 | 12 | 1693 | |||
XM_006724001.1 | XP_006724064.1 | 12 | 1755 | |||
XM_017028341.1 | XP_016883830.1 | 12 | 1750 | |||
GenBank mRNA | AB208909.1 | - | 11 | 3561 | ||
AK095992.1 | - | 12 | 1693 | |||
AK097216.1 | - | 3 | 905 | |||
AK128344.1 | - | 12 | 1563 | |||
AK222505.1 | - | 12 | 1595 | |||
AK303488.1 | - | 12 | 1606 | |||
BC005861.2 | - | 11 | 1549 | 3532902 | ||
BC021077.1 | - | 11 | 1472 | |||
L78790.1 | - | 11 | 1385 | |||
M15395.1 | - | 12 | 1577 | |||
Y00057.1 | - | 11 | 1481 |