Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r14d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028104.4 | NP_082380.1 | 1, 2 | 320 | ||
GenBank mRNA | AF408400.2 | - | 1, 2 | 319 | ||
AK008348.1 | - | 1, 2 | 278 | |||
BC024547.1 | - | 1, 2 | 319 | 4989982 |