Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zswim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027964.2 | NP_082240.1 | 3, 4 | 328 | ||
GenBank mRNA | AK006367.1 | - | 3, 4 | 329 | ||
AK017097.1 | - | 3, 4 | 329 | |||
BC120837.1 | - | 3, 4 | 308 | 40129760 | ||
BC144860.1 | - | 3, 4 | 308 |