Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166413.1 | NP_001159885.1 | 5 | 572 | ||
NM_028027.3 | NP_082303.2 | 5 | 634 | |||
GenBank mRNA | AF487515.1 | - | 5 | 600 | ||
AK010452.1 | - | 5 | 600 | |||
AK158684.1 | - | 5 | 572 | |||
AK210287.1 | - | 2 | 129 | |||
BC041113.1 | - | 5 | 616 | |||
CF724487.1 | - | 5 | 519 |