Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000229.1 | NP_000220.1 | 5 | 688 | ||
GenBank mRNA | BC014781.1 | - | 5 | 678 | 4155094 | |
BT009748.1 | - | 5 | 677 | |||
M12625.1 | - | 5 | 944 | |||
M26268.1 | - | 5 | 630 | |||
X06537.1 | - | 5 | 780 |