Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028049.2 | NP_082325.2 | 5 | 704 | ||
GenBank mRNA | AK002745.1 | - | 5 | 584 | ||
AK005473.1 | - | 4 | 532 | |||
AK182889.1 | - | 1 | 139 | |||
AK188724.1 | - | 1 | 139 | |||
AK207864.1 | - | 1 | 139 | |||
AK213133.1 | - | 1 | 139 | |||
AK219248.1 | - | 1 | 139 | |||
BC018273.1 | - | 5 | 644 | 4458296 |