Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000231.2 | NP_000222.1 | 8 | 1553 | ||
XM_005266505.2 | XP_005266562.1 | 9 | 1646 | |||
XM_006719861.3 | XP_006719924.1 | 8 | 1737 | |||
GenBank mRNA | U34976.1 | - | 9 | 1522 |