Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sulf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252578.1 | NP_001239507.1 | 18 | 2895 | ||
NM_001252579.1 | NP_001239508.1 | 18 | 2741 | |||
NM_028072.5 | NP_082348.2 | 18 | 2737 | |||
GenBank mRNA | AK008108.1 | - | 9 | 1000 | ||
AK028874.1 | - | 18 | 2652 | |||
AK034712.1 | - | 18 | 2722 | |||
AK036685.1 | - | 18 | 2673 | |||
AK049170.1 | - | 13 | 1652 | |||
AK081643.1 | - | 18 | 2695 | |||
AK129316.1 | - | 18 | 2895 | |||
AK133336.1 | - | 2711 | ||||
AK165183.1 | - | 18 | 2649 | |||
AY101177.1 | - | 17 | 2556 | |||
BC027238.1 | - | 7 | 711 | |||
BC062900.1 | - | 18 | 2693 | |||
BC141086.1 | - | 17 | 2490 | 9055698 |