Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028041.2 | NP_082317.1 | 18 | 2220 | ||
GenBank mRNA | AF319547.1 | - | 18 | 2580 | ||
AK010484.1 | - | 15 | 1801 | |||
AK028274.1 | - | 18 | 2235 | |||
AK031534.1 | - | 18 | 2230 | |||
AK031679.1 | - | 18 | 2238 | |||
AK147715.1 | - | 18 | 2236 | |||
AK152989.1 | - | 18 | 2236 | |||
AK155138.1 | - | 18 | 2232 | |||
AK167603.1 | - | 18 | 2234 | |||
AK168455.1 | - | 18 | 2234 | |||
AK169255.1 | - | 18 | 2233 | |||
AK170967.1 | - | 18 | 2232 | |||
BC021900.1 | - | 9 | 1225 | |||
BC043699.1 | - | 18 | 2220 | 5344158 | ||
BC055026.1 | - | 14 | 1663 | |||
BC062920.1 | - | 16 | 3556 | |||
BC071251.1 | - | 12 | 1517 |