Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028064.2 | NP_082340.1 | 7 | 1378 | ||
GenBank mRNA | AK005535.1 | - | 7 | 1418 | ||
AK146977.1 | - | 7 | 1419 | |||
AK147107.1 | - | 7 | 1397 | |||
AK154089.1 | - | 7 | 1897 | |||
AK155467.1 | - | 7 | 1897 | |||
BC023498.1 | - | 7 | 1378 | 5011088 |