Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028127.3 | NP_082403.2 | 14 | 3215 | ||
GenBank mRNA | AK011469.1 | - | 1 | 579 | ||
BC019939.1 | - | 1 | 853 | |||
BC053929.1 | - | 14 | 3215 | 6389695 |