Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mms19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028152.3 | NP_082428.1 | 4 | 334 | ||
GenBank mRNA | AF319949.1 | - | 4 | 328 | ||
AK010326.1 | - | 4 | 338 | |||
AK011754.1 | - | 4 | 328 | |||
AK028317.1 | - | 4 | 334 | |||
AK030619.1 | - | 4 | 335 | |||
AK036983.1 | - | 4 | 328 | |||
AK044710.1 | - | 1 | 261 | |||
AK050265.1 | - | 1 | 270 | |||
AK078688.1 | - | 4 | 329 | |||
AK087633.1 | - | 4 | 335 | |||
BC018614.1 | - | 4 | 312 | |||
BC050817.1 | - | 4 | 314 | 6332214 |