Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luc7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025881.3 | NP_080157.1 | 7 | 837 | ||
NM_028190.3 | NP_082466.2 | 7 | 837 | |||
GenBank mRNA | AK010539.1 | - | 7 | 789 | ||
AK017655.1 | - | 7 | 797 | |||
AK160293.1 | - | 7 | 838 | |||
BC019194.1 | - | 8 | 830 | |||
BC034135.1 | - | 6 | 1003 | 5003664 | ||
BC055875.1 | - | 7 | 1820 | |||
BQ572015.1 | - | 5 | 489 | |||
CF951283.1 | - | 3 | 233 |