Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028325.3 | NP_082601.1 | 5 | 1139 | ||
GenBank mRNA | AK012833.1 | - | 5 | 1056 | ||
AY466375.1 | - | 2 | 905 | |||
BC049071.1 | - | 4 | 1035 | 5701407 | ||
BC055714.1 | - | 4 | 1006 | 6415226 |