Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163746.1 | NP_001157218.1 | 12 | 2944 | ||
NM_028352.4 | NP_082628.3 | 13 | 3067 | |||
GenBank mRNA | AK028066.1 | - | 13 | 3057 | ||
AK036561.1 | - | 13 | 3053 | |||
BC138700.1 | - | 13 | 2992 | 8861722 |