Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dppa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018002.1 | NP_001018002.1 | 3 | 247 | ||
NM_028610.2 | NP_082886.2 | 3 | 247 | |||
GenBank mRNA | AF490348.1 | - | 3 | 164 | ||
AK010753.1 | - | 3 | 215 | |||
AK033197.1 | - | 3 | 248 | |||
BY731814.1 | - | 3 | 213 |