Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esyt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028731.5 | NP_083007.2 | 15 | 1656 | ||
GenBank mRNA | AK014813.1 | - | 17 | 1766 | ||
AK044360.1 | - | 15 | 1658 | |||
AK157352.1 | - | 15 | 1625 | |||
BC040392.1 | - | 5 | 339 | |||
BC052440.1 | - | 5 | 347 | |||
BC054797.1 | - | 2 | 309 | |||
BC059230.1 | - | 5 | 347 | |||
BC138937.1 | - | 15 | 1565 | 8861959 |