Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028477.2 | NP_082753.1 | 5, 6 | 603 | ||
GenBank mRNA | AF525158.1 | - | 5, 6 | 560 | ||
AK005485.1 | - | 5, 6 | 604 | |||
BC109185.1 | - | 5, 6 | 602 | 40050251 | ||
BC109186.1 | - | 5, 6 | 602 | 40050253 |