Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028627.2 | NP_082903.2 | 18 | 3761 | ||
GenBank mRNA | AK003542.1 | - | 2 | 737 | ||
AK158685.1 | - | 8 | 1569 | |||
AK158851.1 | - | 8 | 1693 | |||
AK220438.1 | - | 13 | 3879 | |||
BC043135.1 | - | 14 | 2591 | |||
BC057353.1 | - | 18 | 3763 | |||
BC058352.1 | - | 4 | 1136 | |||
BC069847.1 | - | 1 | 458 |