Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cst6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028623.5 | NP_082899.1 | 3 | 1983 | ||
GenBank mRNA | AK078116.1 | - | 3 | 1983 |